Assay Details
| Gene Symbol: APELA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308250.1 | NP_001295179.1 | 1 - 2 | 120 | 72 | |
| XM_015285308.2 | XP_015140794.1 | 2 - 3 | 297 | 72 | ||
| GenBank mRNA | BG712694.1 | - | 1 - 2 | 120 | 72 | |
| KX017223.1 | - | 1 - 2 | 72 | |||
More Information
Set Membership: |
|

APELA
Genomic Map