Assay Details
| Gene Symbol: TCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310087.1 | NP_001297016.1 | 8 - 9 | 993 | 61 | |
| XM_015288984.2 | XP_015144470.1 | 6 - 7 | 13095 | 61 | ||
| GenBank mRNA | CR385782.1 | - | 8 - 9 | 993 | 61 | |
| CR386579.1 | - | 5 - 6 | 607 | 61 | ||
More Information
Set Membership: |
|

TCTN3
Genomic Map