Assay Details
| Gene Symbol: CSF2RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312662.1 | NP_001299591.1 | 1 - 2 | 141 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001312662.1 transcripts.
Set Membership: |
|

CSF2RA
Genomic Map