Assay Details
| Gene Symbol: LOC776816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308613.2 | NP_001295542.1 | 3 - 4 | 452 | 89 | |
| GenBank mRNA | AJ719332.1 | - | 3 - 4 | 448 | 89 | |
| AJ719605.1 | - | 3 - 4 | 448 | 89 | ||
More Information
Set Membership: |
|

LOC776816
Genomic Map