Assay Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318990.1 | NP_001305919.1 | 3 - 4 | 841 | 95 | |
| XM_004942644.3 | XP_004942701.2 | 3 - 4 | 919 | 95 | ||
| GenBank mRNA | JQ736790.1 | - | 3 - 4 | 841 | 95 | |
More Information
Set Membership: |
|

KLF7
Genomic Map