Assay Details
| Gene Symbol: UQCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302149.1 | NP_001289078.1 | 2 | 318 | 95 | |
| GenBank mRNA | BX933152.1 | - | 1 | 891 | 95 | |
| BX935769.2 | - | 2 | 296 | 95 | ||
| CF249663.1 | - | 2 | 320 | 95 | ||
More Information
Set Membership: |
|

UQCR10
Genomic Map