Assay Details
| Gene Symbol: COA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302185.1 | NP_001289114.1 | 3 | 815 | 79 | |
| GenBank mRNA | BX930638.1 | - | 2 | 79 | ||
| BX934527.2 | - | 3 | 815 | 79 | ||
| CR385316.1 | - | 4 | 79 | |||
| CR385317.1 | - | 3 | 79 | |||
| CR406778.1 | - | 4 | 79 | |||
More Information
Set Membership: |
|

COA5
Genomic Map