Assay Details
| Gene Symbol: CATH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001605.3 | NP_001001605.1 | 1 | 225 | 83 | |
| GenBank mRNA | BX936022.1 | - | 1 | 225 | 83 | |
| Gene Symbol: CATH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025146986.1 | XP_025002754.1 | 4 | 3964 | 83 | |
| GenBank mRNA | DQ092351.1 | - | 1 | 129 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001001605.3, XM_025146986.1 transcripts.
Set Membership: |
|

CATH1
Genomic Map