Assay Details
| Gene Symbol: GLRX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008472.1 | NP_001008472.1 | 2 | 2220 | 109 | |
| GenBank mRNA | AJ720261.1 | - | 2 | 2220 | 109 | |
More Information
Set Membership: |
|

GLRX5
Genomic Map