Assay Details
| Gene Symbol: RNF139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030955.1 | NP_001026126.1 | 2 | 1624 | 89 | |
| GenBank mRNA | AJ720916.1 | - | 2 | 1705 | 89 | |
More Information
Set Membership: |
|

RNF139
Genomic Map