Assay Details
| Gene Symbol: LDLRAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031013.2 | NP_001026184.2 | 4 | 918 | 64 | |
| XM_015282455.2 | XP_015137941.1 | 6 | 1630 | 64 | ||
| XM_015282456.2 | XP_015137942.1 | 4 | 1309 | 64 | ||
| XM_015282457.2 | XP_015137943.1 | 3 | 1255 | 64 | ||
| XM_015282459.2 | XP_015137945.1 | 4 | 934 | 64 | ||
| XM_015282461.2 | XP_015137947.1 | 4 | 910 | 64 | ||
| XM_015282462.2 | XP_015137948.1 | 4 | 886 | 64 | ||
| XM_025147299.1 | XP_025003067.1 | 4 | 911 | 64 | ||
| GenBank mRNA | AJ720368.1 | - | 3 | 2126 | 64 | |
More Information
Set Membership: |
|

LDLRAD4
Genomic Map