Assay Details
| Gene Symbol: TWIST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103214.1 | NP_001096684.1 | 2 | 1018 | 84 | |
| GenBank mRNA | BX929837.1 | - | 2 | 552 | 84 | |
| BX929903.1 | - | 2 | 1020 | 84 | ||
More Information
Set Membership: |
|

TWIST3
Genomic Map