Assay Details
| Gene Symbol: GPER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162405.1 | NP_001155877.1 | 2 | 720 | 94 | |
| XM_004945130.3 | XP_004945187.2 | 4 | 8436 | 94 | ||
| XM_015294159.2 | XP_015149645.1 | 5 | 7937 | 94 | ||
| GenBank mRNA | FJ883174.1 | - | 2 | 720 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015294159.2 transcripts.
Set Membership: |
|

GPER1
Genomic Map