Assay Details
| Gene Symbol: UQCR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190802.1 | NP_001177731.1 | 3 | 325 | 63 | |
| GenBank mRNA | BU216138.1 | - | 3 | 326 | 63 | |
| BX935808.1 | - | 3 | 318 | 63 | ||
| BX936178.1 | - | 3 | 329 | 63 | ||
More Information
Set Membership: |
|

UQCR11
Genomic Map