Assay Details
| Gene Symbol: BBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277511.2 | NP_001264440.1 | 3 | 525 | 129 | |
| GenBank mRNA | CN383547.1 | - | 3 | 521 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BBIP1| Gene Symbol: BBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277511.2 | NP_001264440.1 | 3 | 525 | 129 | |
| GenBank mRNA | CN383547.1 | - | 3 | 521 | 129 | |
Set Membership: |
|