Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277615.3 | NP_001264544.1 | 4 | 320 | 147 | |
| XM_015292510.2 | XP_015147996.2 | 4 | 727 | 147 | ||
| GenBank mRNA | BX950673.2 | - | 3 | 423 | 147 | |
More Information
Set Membership: |
|

DYNLRB2
Genomic Map