Assay Details
| Gene Symbol: LOC107050456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX950841.1 | - | 3 - 5 | 506 | 52 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107050456| Gene Symbol: LOC107050456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX950841.1 | - | 3 - 5 | 506 | 52 | |