Assay Details
| Gene Symbol: GNAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232444.4 | XP_001232445.1 | 2 - 3 | 1764 | 82 | |
| XM_015275439.2 | XP_015130925.1 | 3 - 4 | 1863 | 82 | ||
| GenBank mRNA | CR352420.1 | - | 1 - 2 | 875 | 82 | |
More Information
Set Membership: |
|

GNAZ
Genomic Map