Assay Details
| Gene Symbol: RCOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287816.2 | XP_015143302.1 | 10 - 11 | 1126 | 62 | |
| XM_025150951.1 | XP_025006719.1 | 9 - 10 | 864 | 62 | ||
| GenBank mRNA | CR352626.1 | - | 10 - 11 | 1073 | 62 | |
More Information
Set Membership: |
|

RCOR1
Genomic Map