Assay Details
| Gene Symbol: MRPS36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234564.5 | XP_001234565.2 | 3 - 4 | 349 | 88 | |
| GenBank mRNA | BX929351.1 | - | 3 - 4 | 317 | 88 | |
More Information
Set Membership: |
|

MRPS36
Genomic Map