Assay Details
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015276695.2 | XP_015132181.1 | 2 - 3 | 236 | 145 | |
XM_015276696.2 | XP_015132182.1 | 2 - 3 | 236 | 145 | ||
GenBank mRNA | BX929445.1 | - | 2 - 3 | 226 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
