Assay Details
| Gene Symbol: CA15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415218.6 | XP_415218.3 | 3 - 4 | 452 | 83 | |
| GenBank mRNA | BX929589.1 | - | 3 - 4 | 362 | 83 | |
| CR352958.1 | - | 3 - 4 | 387 | 83 | ||
More Information
Set Membership: |
|

CA15L
Genomic Map