Assay Details
| Gene Symbol: CA15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415218.6 | XP_415218.3 | 5 - 6 | 711 | 67 | |
| GenBank mRNA | BX929589.1 | - | 5 - 6 | 621 | 67 | |
| CR352958.1 | - | 5 - 6 | 646 | 67 | ||
More Information
Set Membership: |
|

CA15L
Genomic Map