Assay Details
| Gene Symbol: KLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285673.2 | XP_015141159.1 | 5 - 6 | 909 | 69 | |
| XM_015285674.2 | XP_015141160.1 | 5 - 6 | 3356 | 69 | ||
| XM_015285675.2 | XP_015141161.1 | 4 - 5 | 930 | 69 | ||
| XM_015285676.2 | XP_015141162.1 | 3 - 4 | 749 | 69 | ||
| XM_025149970.1 | XP_025005738.1 | 5 - 6 | 919 | 69 | ||
| GenBank mRNA | BX929685.2 | - | 2 - 3 | 505 | 69 | |
| JX673910.1 | - | 3 - 4 | 665 | 69 | ||
More Information
Set Membership: |
|

KLF3
Genomic Map