Assay Details
| Gene Symbol: PLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234074.5 | XP_001234075.5 | 2 - 3 | 196 | 63 | |
| GenBank mRNA | BX929686.1 | - | 1 - 2 | 32 | 63 | |
More Information
Set Membership: |
|

PLPP5
Genomic Map