Assay Details
| Gene Symbol: AS3MT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421735.6 | XP_421735.3 | 7 - 8 | 676 | 68 | |
| GenBank mRNA | BX929700.2 | - | 5 - 6 | 576 | 68 | |
| BX934758.1 | - | 5 - 6 | 678 | 68 | ||
More Information
Set Membership: |
|

AS3MT
Genomic Map