Assay Details
| Gene Symbol: LOC100858626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290087.2 | XP_015145573.1 | 2 - 3 | 187 | 75 | |
| XM_025152440.1 | XP_025008208.1 | 2 - 3 | 539 | 75 | ||
| GenBank mRNA | BX929761.1 | - | 1 - 2 | 50 | 75 | |
| BX935455.2 | - | 2 - 3 | 196 | 75 | ||
More Information
Set Membership: |
|

LOC100858626
Genomic Map