Assay Details
| Gene Symbol: LOC100858626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290087.2 | XP_015145573.1 | 6 - 7 | 650 | 72 | |
| XM_025152440.1 | XP_025008208.1 | 6 - 7 | 1002 | 72 | ||
| GenBank mRNA | BX929761.1 | - | 5 - 6 | 513 | 72 | |
| BX935455.2 | - | 6 - 7 | 659 | 72 | ||
More Information
Set Membership: |
|

LOC100858626
Genomic Map