Assay Details
| Gene Symbol: IMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285172.2 | XP_015140658.1 | 1 | 1097 | 98 | |
| GenBank mRNA | BX929780.1 | - | 1 | 626 | 98 | |
| CR387273.1 | - | 1 | 616 | 98 | ||
More Information
Set Membership: |
|

IMP3
Genomic Map