Assay Details
| Gene Symbol: PTP4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297841.2 | XP_015153327.1 | 3 - 4 | 613 | 77 | |
| XM_015297845.2 | XP_015153331.1 | 2 - 3 | 742 | 77 | ||
| XM_025143086.1 | XP_024998854.1 | 2 - 3 | 737 | 77 | ||
| GenBank mRNA | BX929867.1 | - | 3 - 4 | 784 | 77 | |
| BX931523.2 | - | 1 - 2 | 337 | 77 | ||
| BX931762.2 | - | 1 - 2 | 643 | 77 | ||
| BX935249.1 | - | 1 - 2 | 549 | 77 | ||
More Information
Set Membership: |
|

PTP4A2
Genomic Map