Assay Details
| Gene Symbol: SPAG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296077.1 | XP_015151563.1 | 19 - 20 | 106 | ||
| XM_025142076.1 | XP_024997844.1 | 19 - 20 | 106 | |||
| GenBank mRNA | CR352895.1 | - | 1 - 2 | 76 | 106 | |
More Information
Set Membership: |
|

SPAG5
Genomic Map