Assay Details
| Gene Symbol: EXTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297760.2 | XP_015153246.1 | 7 - 8 | 2158 | 92 | |
| GenBank mRNA | CR353193.1 | - | 1 - 2 | 155 | 92 | |
More Information
Set Membership: |
|

EXTL1
Genomic Map