Assay Details
| Gene Symbol: RASSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292789.2 | XP_015148275.1 | 4 - 5 | 1011 | 90 | |
| XM_015292790.2 | XP_015148276.1 | 5 - 6 | 896 | 90 | ||
| GenBank mRNA | CR353234.1 | - | 4 - 5 | 589 | 90 | |
More Information
Set Membership: |
|

RASSF1
Genomic Map