Assay Details
| Gene Symbol: ARL8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293177.2 | XP_015148663.1 | 5 - 6 | 367 | 84 | |
| XM_015293178.2 | XP_015148664.1 | 5 - 6 | 349 | 84 | ||
| XM_414440.6 | XP_414440.3 | 5 - 6 | 516 | 84 | ||
| GenBank mRNA | CR353485.1 | - | 5 - 6 | 559 | 84 | |
More Information
Set Membership: |
|

ARL8B
Genomic Map