Assay Details
| Gene Symbol: NEFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415310.5 | XP_415310.4 | 4 | 3424 | 88 | |
| GenBank mRNA | CR353544.1 | - | 1 | 925 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEFH| Gene Symbol: NEFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415310.5 | XP_415310.4 | 4 | 3424 | 88 | |
| GenBank mRNA | CR353544.1 | - | 1 | 925 | 88 | |
Set Membership: |
|