Assay Details
| Gene Symbol: RPH3AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235227.4 | XP_001235228.1 | 9 | 2436 | 108 | |
| XM_004946702.3 | XP_004946759.1 | 8 | 6023 | 108 | ||
| XM_004946703.3 | XP_004946760.1 | 9 | 2465 | 108 | ||
| XM_004946704.3 | XP_004946761.1 | 7 | 5937 | 108 | ||
| XM_004946705.3 | XP_004946762.1 | 9 | 5896 | 108 | ||
| XM_004946706.3 | XP_004946763.1 | 9 | 2448 | 108 | ||
| XM_004946707.3 | XP_004946764.1 | 9 | 2438 | 108 | ||
| XM_015296119.2 | XP_015151605.1 | 9 | 5211 | 108 | ||
| XM_025142000.1 | XP_024997768.1 | 9 | 2378 | 108 | ||
| XM_025142001.1 | XP_024997769.1 | 9 | 4252 | 108 | ||
| GenBank mRNA | CR353553.1 | - | 1 | 1325 | 108 | |
More Information
Set Membership: |
|

RPH3AL
Genomic Map