Assay Details
| Gene Symbol: CBSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300878.1 | XP_015156364.1 | 3 - 4 | 447 | 77 | |
| XM_015300883.2 | XP_015156369.1 | 3 - 4 | 353 | 77 | ||
| XM_416752.5 | XP_416752.3 | 3 - 4 | 446 | 77 | ||
| GenBank mRNA | CR353850.1 | - | 3 - 4 | 348 | 77 | |
More Information
Set Membership: |
|

CBSL
Genomic Map