Assay Details
| Gene Symbol: CBSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300878.1 | XP_015156364.1 | 6 - 7 | 759 | 97 | |
| XM_015300883.2 | XP_015156369.1 | 6 - 7 | 665 | 97 | ||
| XM_416752.5 | XP_416752.3 | 6 - 7 | 758 | 97 | ||
| GenBank mRNA | CR353850.1 | - | 6 - 7 | 660 | 97 | |
More Information
Set Membership: |
|

CBSL
Genomic Map