Assay Details
| Gene Symbol: NAA30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151311.1 | XP_025007079.1 | 1 | 602 | 112 | |
| GenBank mRNA | CR353964.1 | - | 1 | 221 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAA30| Gene Symbol: NAA30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151311.1 | XP_025007079.1 | 1 | 602 | 112 | |
| GenBank mRNA | CR353964.1 | - | 1 | 221 | 112 | |
Set Membership: |
|