Assay Details
| Gene Symbol: MMAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155672.1 | XP_025011440.1 | 4 - 5 | 361 | 69 | |
| GenBank mRNA | CR354091.1 | - | 1 - 2 | 65 | 69 | |
| CR390775.1 | - | 2 - 3 | 221 | 69 | ||
More Information
Set Membership: |
|

MMAB
Genomic Map