Assay Details
| Gene Symbol: CYB5D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415743.6 | XP_415743.1 | 3 - 4 | 594 | 92 | |
| GenBank mRNA | CR385216.1 | - | 1 - 2 | 118 | 92 | |
More Information
Set Membership: |
|

CYB5D2
Genomic Map