Assay Details
| Gene Symbol: PUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945515.3 | XP_004945572.2 | 2 - 3 | 354 | 99 | |
| XM_415090.6 | XP_415090.1 | 2 - 3 | 404 | 99 | ||
| GenBank mRNA | CR385347.1 | - | 1 - 2 | 81 | 99 | |
More Information
Set Membership: |
|

PUS1
Genomic Map