Assay Details
| Gene Symbol: TSPAN15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142799.1 | XP_024998567.1 | 6 - 7 | 2790 | 142 | |
| GenBank mRNA | CR385382.1 | - | 2 - 3 | 655 | 142 | |
More Information
Set Membership: |
|

TSPAN15L
Genomic Map