Assay Details
| Gene Symbol: TSPAN15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142799.1 | XP_024998567.1 | 10 - 11 | 3130 | 115 | |
| XM_025142800.1 | XP_024998568.1 | 8 - 9 | 3054 | 115 | ||
| GenBank mRNA | CR385382.1 | - | 6 - 7 | 995 | 115 | |
More Information
Set Membership: |
|

TSPAN15L
Genomic Map