Assay Details
| Gene Symbol: PIK3R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426652.6 | XP_426652.3 | 16 | 7928 | 101 | |
| GenBank mRNA | CR385542.1 | - | 1 | 817 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIK3R3| Gene Symbol: PIK3R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426652.6 | XP_426652.3 | 16 | 7928 | 101 | |
| GenBank mRNA | CR385542.1 | - | 1 | 817 | 101 | |
Set Membership: |
|