Assay Details
| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004949016.3 | XP_004949073.1 | 5 | 5920 | 76 | |
| XM_004949017.3 | XP_004949074.1 | 7 | 6143 | 76 | ||
| XM_015277408.2 | XP_015132894.1 | 7 | 6153 | 76 | ||
| XM_015277409.2 | XP_015132895.1 | 7 | 6622 | 76 | ||
| XM_025144987.1 | XP_025000755.1 | 5 | 7995 | 76 | ||
| XM_429131.6 | XP_429131.3 | 7 | 6087 | 76 | ||
| GenBank mRNA | CR385845.1 | - | 1 | 616 | 76 | |
More Information
Set Membership: |
|

ZBTB7C
Genomic Map