Assay Details
| Gene Symbol: PCASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642904.4 | XP_003642952.4 | 17 | 4748 | 101 | |
| XM_004949002.3 | XP_004949059.3 | 17 | 4751 | 101 | ||
| XM_004949003.3 | XP_004949060.3 | 16 | 4718 | 101 | ||
| XM_004949004.3 | XP_004949061.3 | 16 | 4715 | 101 | ||
| XM_025144928.1 | XP_025000696.1 | 13 | 4451 | 101 | ||
| GenBank mRNA | CR385977.1 | - | 1 | 625 | 101 | |
More Information
Set Membership: |
|

PCASP1
Genomic Map