Assay Details
| Gene Symbol: PXYLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277036.2 | XP_015132522.1 | 5 | 2712 | 146 | |
| GenBank mRNA | CR386084.1 | - | 1 | 1022 | 147 | |
More Information
Set Membership: |
|

PXYLP1
Genomic Map