Assay Details
| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937339.3 | XP_004937396.1 | 13 | 9895 | 88 | |
| XM_429151.6 | XP_429151.3 | 13 | 9420 | 88 | ||
| GenBank mRNA | CR386267.1 | - | 1 | 819 | 88 | |
More Information
Set Membership: |
|

SV2C
Genomic Map