Assay Details
| Gene Symbol: CABLES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278006.2 | XP_015133492.2 | 10 | 7664 | 102 | |
| XM_025147894.1 | XP_025003662.1 | 9 | 7586 | 102 | ||
| XM_025147895.1 | XP_025003663.1 | 10 | 2945 | 102 | ||
| GenBank mRNA | CR386290.1 | - | 1 | 419 | 102 | |
More Information
Set Membership: |
|

CABLES1
Genomic Map